3,6-Difluoro-2-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

29247-500MG 29247-1G 29247-5G
77337-396EA 414.06 USD
77337-396 77348-814 77373-076
3,6-Difluoro-2-methylbenzyl alcohol ≥97%
3,6-Difluoro-2-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₈F₂O
MW: 158.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16250141
CAS Number: 1378906-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR