4-Benzyloxy-2-(trifluoromethyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

29214-500MG 29214-1G 29214-5G 29214-25G
77325-002EA 175.1 USD
77325-002 77346-478 77361-336 77384-562
4-Benzyloxy-2-(trifluoromethyl)benzoic acid ≥95%
4-Benzyloxy-2-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₁F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD15528836
CAS Number: 790695-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR