4-Benzyloxy-2-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

29212-500MG 29212-1G 29212-5G
77336-326EA 387.28 USD
77336-326 77348-808 77373-072
4-Benzyloxy-2-(trifluoromethyl)benzaldehyde ≥97%
4-Benzyloxy-2-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₁F₃O₂
MW: 280.242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15528835
CAS Number: 678164-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR