3-(Benzyloxy)-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

29202-500MG 29202-1G 29202-5G 29202-25G
77325-000EA 229.69 USD
77325-000 77348-806 77361-334 77384-560
3-(Benzyloxy)-2-methoxybenzaldehyde ≥97%
3-(Benzyloxy)-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22041844
CAS Number: 273200-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR