4-Benzyloxy-2-(trifluoromethoxy)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

29198-500MG 29198-1G 29198-5G 29198-25G
77324-998EA 181.28 USD
77324-998 77346-788 77361-332 77384-558
4-Benzyloxy-2-(trifluoromethoxy)phenylboronic acid pinacol ester ≥97%
4-Benzyloxy-2-(trifluoromethoxy)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₀H₂₂BF₃O₄
Storage Temperature: Ambient
MDL Number: MFCD28806023
CAS Number: 2121511-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR