2-Methyl-4-(methylsulfanyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

29138-500MG 29138-1G 29138-5G 29138-25G
77324-994EA 233.81 USD
77324-994 77348-802 77361-330 77384-556
2-Methyl-4-(methylsulfanyl)benzaldehyde ≥95%
2-Methyl-4-(methylsulfanyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀OS
Storage Temperature: Ambient
MDL Number: MFCD18887986
CAS Number: 956612-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR