2-Methoxy-5-trifluoromethoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(2-Methoxy-5-(trifluoromethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

29126-1G 29126-5G 29126-25G
77342-112EA 114.33 USD
77342-112 77361-326 77373-066
2-Methoxy-5-trifluoromethoxyphenylboronic acid pinacol ester ≥97%
2-Methoxy-5-trifluoromethoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BF₃O₄
MW: 318.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22415050
CAS Number: 1688666-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR