Ethyl 2,3-dichloroisonicotinate ≥95%

Supplier: AOBChem USA

Warning

29109-500MG 29109-1G 29109-5G 29109-25G
77324-990EA 99.91 USD
77324-990 77342-762 77361-324 77373-062
Ethyl 2,3-dichloroisonicotinate ≥95%
Ethyl 2,3-dichloroisonicotinate

Precaución: for Research Use Only

Formula: C₈H₇Cl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD11656325
CAS Number: 1092353-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR