3-Bromo-4-methoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

29085-500MG 29085-1G 29085-5G 29085-25G
77324-986EA 284.28 USD
77324-986 77348-788 77361-318 77384-550
3-Bromo-4-methoxyphenylboronic acid pinacol ester ≥97%
3-Bromo-4-methoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BBrO₃
MW: 312.995 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28806019
CAS Number: 1394130-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR