(3-Bromo-5-cyanophenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

29075-500MG 29075-1G 29075-5G
77340-188EA 822.97 USD
77340-188 77348-784 77373-052
(3-Bromo-5-cyanophenyl)boronic acid ≥95%
(3-Bromo-5-cyanophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₅BBrNO₂
MW: 225.835 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22544488
CAS Number: 1658464-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR