1-Bromo-3-benzyloxy-4,5-difluorobenzene ≥95%

Supplier: AOBChem USA

Warning

29063-500MG 29063-1G 29063-5G 29063-25G
77324-980EA 112.27 USD
77324-980 77343-300 77361-312 77373-050
1-Bromo-3-benzyloxy-4,5-difluorobenzene ≥95%
1-Bromo-3-benzyloxy-4,5-difluorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉BrF₂O
MW: 299.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17169369
CAS Number: 1035155-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR