4-Isopropoxy-2-methylbenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
28999-500MG
28999-1G
28999-5G
28999-25G
77324-978EA
160.68
USD
77324-978
77345-760
77361-310
77384-544
4-Isopropoxy-2-methylbenzoic acid ≥97%
4-Isopropoxy-2-methylbenzoic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₄O₃ Storage Temperature: Ambient |
MDL Number:
MFCD19309246 CAS Number: 111220-35-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...