1-Bromo-2,3-difluoro-4-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

28991-500MG 28991-1G 28991-5G 28991-25G
77324-976EA 225.57 USD
77324-976 77348-780 77361-308 77384-542
1-Bromo-2,3-difluoro-4-isopropoxybenzene ≥97%
1-Bromo-2,3-difluoro-4-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉BrF₂O
MW: 251.068 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29043843
CAS Number: 1881330-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR