5-Cyclopropyl-2-fluoropyridine-3-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

28785-500MG 28785-1G 28785-5G 28785-25G
77324-970EA 182.31 USD
77324-970 77346-904 77361-302 77384-538
5-Cyclopropyl-2-fluoropyridine-3-carboxylic acid ≥95%
5-Cyclopropyl-2-fluoropyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD28388513
CAS Number: 1402664-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR