Potassium (5-hydroxy-2-methylphenyl)trifluoroborate ≥97%

Supplier: AOBChem USA

Warning

28713-500MG 28713-1G 28713-5G 28713-25G
77324-966EA 233.81 USD
77324-966 77348-776 77361-298 77384-534
Potassium (5-hydroxy-2-methylphenyl)trifluoroborate ≥97%
Potassium (5-hydroxy-2-methylphenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₇H₇BF₃KO
MW: 214.034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28388510
CAS Number: 2149596-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR