Potassium (3-hydroxy-4-methylphenyl)trifluoroborate ≥95%

Supplier: AOBChem USA

Warning

28712-500MG 28712-1G 28712-5G 28712-25G
77324-964EA 136.99 USD
77324-964 77344-580 77361-296 77384-532
Potassium (3-hydroxy-4-methylphenyl)trifluoroborate ≥95%
Potassium (3-hydroxy-4-methylphenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₇H₇BF₃KO
MW: 214.034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09993092
CAS Number: 2149596-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR