Potassium (2-acetylphenyl)trifluoroborate ≥95%

Supplier: AOBChem USA

Warning

28708-1G 28708-5G 28708-25G
77341-068EA 69.01 USD
77341-068 77361-294 77373-046
Potassium (2-acetylphenyl)trifluoroborate ≥95%
Potassium (2-acetylphenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₈H₇BF₃KO
MW: 226.045 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09993258
CAS Number: 1258323-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR