Borate(1-), trifluoro-1H-indazol-4-yl-, potassium (1:1), (T-4)- ≥95%

Supplier: AOBChem USA

Warning

28661-500MG 28661-1G 28661-5G 28661-25G
77324-962EA 112.27 USD
77324-962 77343-326 77361-292 77373-042
Borate(1-), trifluoro-1H-indazol-4-yl-, potassium (1:1), (T-4)- ≥95%
Borate(1-), trifluoro-1H-indazol-4-yl-, potassium (1:1), (T-4)-

Precaución: for Research Use Only

Formula: C₇H₅BF₃KN₂
MW: 224.033 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09993228
CAS Number: 1542163-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR