2-Phenoxypyridine-4-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

28658-500MG 28658-1G 28658-5G
77336-610EA 390.37 USD
77336-610 77348-774 77373-040
2-Phenoxypyridine-4-boronic acid pinacol ester ≥97%
2-Phenoxypyridine-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₀BNO₃
Storage Temperature: Ambient
MDL Number: MFCD11878169
CAS Number: 2096331-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR