2-Methyl-4-phenoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Methyl-4-phenoxyphenyl)boronic acid

28500-500MG 28500-1G 28500-5G 28500-25G
77324-950EA 116.39 USD
77324-950 77343-410 77361-274 77373-026
2-Methyl-4-phenoxyphenylboronic acid ≥97%
2-Methyl-4-phenoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₃BO₃
Storage Temperature: Ambient
MDL Number: MFCD28009325
CAS Number: 1449001-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR