3-Bromo-5-fluoro-2-methoxybenzonitrile ≥97%

Supplier: AOBChem USA

Warning

28482-500MG 28482-1G 28482-5G 28482-25G
77324-946EA 181.28 USD
77324-946 77346-782 77361-270 77384-522
3-Bromo-5-fluoro-2-methoxybenzonitrile ≥97%
3-Bromo-5-fluoro-2-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrFNO
MW: 230.034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28360692
CAS Number: 1260022-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR