3-Chloro-2-fluoro-N-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

28458-1G 28458-5G 28458-25G
77340-620EA 60.77 USD
77340-620 77361-268 77373-024
3-Chloro-2-fluoro-N-methoxy-N-methylbenzamide ≥97%
3-Chloro-2-fluoro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₉ClFNO₂
Storage Temperature: Ambient
MDL Number: MFCD24020531
CAS Number: 680610-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR