5-n-Propylthiophene-2-boronic acid pinacol este ≥97%

Supplier: AOBChem USA

Warning

28380-500MG 28380-1G 28380-25G 28380-5G
77324-942EA 299.73 USD
77324-942 77348-768 77384-518 77373-022
5-n-Propylthiophene-2-boronic acid pinacol este ≥97%
5-n-Propylthiophene-2-boronic acid pinacol este

Precaución: for Research Use Only

Formula: C₁₃H₂₁BO₂S
MW: 252.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18436112
CAS Number: 1473358-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR