2-Bromo-5-trifluoromethylphenylboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

28357-500MG 28357-1G 28357-5G 28357-25G
77324-936EA 136.99 USD
77324-936 77344-522 77361-260 77384-514
2-Bromo-5-trifluoromethylphenylboronic acid pinacol ester ≥95%
2-Bromo-5-trifluoromethylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BBrF₃O₂
MW: 350.967 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28154574
CAS Number: 2922194-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR