N-(2,2-Dimethoxyethyl)-N-(2-fluorobenzyl)-4-methylbenzenesulfonamide ≥97%

Supplier: AOBChem USA

Warning

28299-500MG 28299-1G 28299-5G 28299-25G
77324-934EA 282.22 USD
77324-934 77348-766 77361-258 77384-512
N-(2,2-Dimethoxyethyl)-N-(2-fluorobenzyl)-4-methylbenzenesulfonamide ≥97%
N-(2,2-Dimethoxyethyl)-N-(2-fluorobenzyl)-4-methylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₈H₂₂FNO₄S
MW: 367.435 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28403410
CAS Number: 1404367-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR