4-Chloro-5-iodo-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

28269-500MG 28269-1G 28269-25G 28269-5G
77324-930EA 299.73 USD
77324-930 77348-762 77384-508 77373-018
4-Chloro-5-iodo-2-methoxypyridine ≥97%
4-Chloro-5-iodo-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅ClINO
MW: 269.467 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18415722
CAS Number: 1261488-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR