2-(6-Bromo-2-fluoro-3-methylphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

28242-1G 28242-5G 28242-25G
77341-080EA 70.04 USD
77341-080 77361-254 77373-016
2-(6-Bromo-2-fluoro-3-methylphenyl)propan-2-ol ≥95%
2-(6-Bromo-2-fluoro-3-methylphenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFO
MW: 247.104 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977280
CAS Number: 1437780-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR