4-Fluorobenzofuran-7-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

28223-500MG 28223-1G 28223-5G
77338-760EA 517.06 USD
77338-760 77348-760 77373-014
4-Fluorobenzofuran-7-carboxylic acid ≥97%
4-Fluorobenzofuran-7-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅FO₃
MW: 180.133 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27938388
CAS Number: 1823956-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR