4-Bromo-6-chloro-3-iodo-2-methylpyridine ≥95%

Supplier: AOBChem USA

Warning

28206-500MG 28206-1G 28206-5G
77324-928EA 346.08 USD
77324-928 77348-758 77373-010
4-Bromo-6-chloro-3-iodo-2-methylpyridine ≥95%
4-Bromo-6-chloro-3-iodo-2-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₄BrClIN
MW: 332.364 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28154571
CAS Number: 1809161-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR