2-Methoxy-4-(methoxymethoxy)-phenylboronic acid, pinacol ester ≥95%

Supplier: AOBChem USA

Warning

28156-500MG 28156-1G 28156-5G 28156-25G
77324-924EA 82.4 USD
77324-924 77342-142 77361-248 77373-006
2-Methoxy-4-(methoxymethoxy)-phenylboronic acid, pinacol ester ≥95%
2-Methoxy-4-(methoxymethoxy)-phenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₃BO₅
Storage Temperature: Ambient
MDL Number: MFCD28154567
CAS Number: 2121513-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR