5,6-Dimethoxy-2-formylpyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5,6-Dimethoxypyridine-2-carboxaldehyde

28128-500MG 28128-1G 28128-5G 28128-25G
77324-922EA 276.04 USD
77324-922 77348-756 77361-246 77384-506
5,6-Dimethoxy-2-formylpyridine ≥97%
5,6-Dimethoxy-2-formylpyridine

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11857645
CAS Number: 106331-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR