2-Iodo-5-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

27984-500MG 27984-1G 27984-5G 27984-25G
77324-916EA 108.15 USD
77324-916 77343-156 77361-242 77373-000
2-Iodo-5-(trifluoromethyl)phenol ≥97%
2-Iodo-5-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₄F₃IO
Storage Temperature: Ambient
MDL Number: MFCD13186736
CAS Number: 102771-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR