3,5-Dimethyl-4-(methylsulfanyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

27937-500MG 27937-1G 27937-5G 27937-25G
77324-908EA 257.5 USD
77324-908 77348-742 77361-234 77384-496
3,5-Dimethyl-4-(methylsulfanyl)benzoic acid ≥95%
3,5-Dimethyl-4-(methylsulfanyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD28977270
CAS Number: 2635937-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR