4-Bromo-1-isopropoxy-2-methoxybenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
27857-500MG
27857-1G
27857-5G
27857-25G
77324-904EA
126.69
USD
77324-904
77344-200
77361-230
77372-998
4-Bromo-1-isopropoxy-2-methoxybenzene ≥97%
4-Bromo-1-isopropoxy-2-methoxybenzene
Precaución: for Research Use Only
Formula:
C₁₀H₁₃BrO₂ Storage Temperature: Ambient |
MDL Number:
MFCD08693145 CAS Number: 138505-27-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...