2,4-Dichloro-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

27841-500MG 27841-1G 27841-5G
77340-096EA 754.99 USD
77340-096 77348-738 77372-996
2,4-Dichloro-5-(trifluoromethyl)benzaldehyde ≥97%
2,4-Dichloro-5-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₃Cl₂F₃O
MW: 243.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34755837
CAS Number: 134099-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR