(4-Bromophenyl)-(2,3-difluorophenyl)methanone ≥97%

Supplier: AOBChem USA

Warning

27835-500MG 27835-1G 27835-5G 27835-25G
77324-902EA 188.49 USD
77324-902 77348-736 77361-226 77384-492
(4-Bromophenyl)-(2,3-difluorophenyl)methanone ≥97%
(4-Bromophenyl)-(2,3-difluorophenyl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₇BrF₂O
MW: 297.095 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18004996
CAS Number: 1282790-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR