8-Iodoimidazo[1,2-a]pyridine ≥97%

Supplier: AOBChem USA

Warning

27821-1G 27821-5G 27821-25G
77340-618EA 60.77 USD
77340-618 77361-224 77372-990
8-Iodoimidazo[1,2-a]pyridine ≥97%
8-Iodoimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₅IN₂
MW: 244.032 g/mol
Storage Temperature: Ambient
CAS Number: 1809158-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR