2-Bromophenyl ethyl sulfide ≥95%

Supplier: AOBChem USA

Warning

27765-500MG 27765-1G 27765-5G 27765-25G
77324-898EA 105.06 USD
77324-898 77343-088 77361-220 77372-986
2-Bromophenyl ethyl sulfide ≥95%
2-Bromophenyl ethyl sulfide

Precaución: for Research Use Only

Formula: C₈H₉BrS
MW: 217.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08460162
CAS Number: 87424-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR