1-Chloro-4-phenoxy-2-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

27713-500MG 27713-1G 27713-5G
77324-894EA 353.29 USD
77324-894 77348-730 77372-984
1-Chloro-4-phenoxy-2-(trifluoromethoxy)benzene ≥97%
1-Chloro-4-phenoxy-2-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₁₃H₈ClF₃O₂
MW: 288.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34755834
CAS Number: 2764728-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR