N-[3-(trifluoromethoxy)phenyl]acetamide ≥97%

Supplier: AOBChem USA

Warning

27707-500MG 27707-1G 27707-5G 27707-25G
77324-892EA 145.23 USD
77324-892 77345-042 77361-216 77384-488
N-[3-(trifluoromethoxy)phenyl]acetamide ≥97%
N-[3-(trifluoromethoxy)phenyl]acetamide

Precaución: for Research Use Only

Formula: C₉H₈F₃NO₂
MW: 219.161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07579066
CAS Number: 1956-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR