3-Fluoro-5-(methylsulfanyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

27659-500MG 27659-1G 27659-5G
77324-890EA 326.51 USD
77324-890 77348-728 77372-980
3-Fluoro-5-(methylsulfanyl)benzonitrile ≥97%
3-Fluoro-5-(methylsulfanyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₆FNS
MW: 167.203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11976043
CAS Number: 453566-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR