4-Bromo-2-chloro-5-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

27642-500MG 27642-1G 27642-25G 27642-5G
77324-888EA 289.43 USD
77324-888 77348-726 77384-486 77372-978
4-Bromo-2-chloro-5-(trifluoromethyl)benzyl alcohol ≥97%
4-Bromo-2-chloro-5-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₅BrClF₃O
MW: 289.477 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28142607
CAS Number: 1809161-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR