6-Methoxy-2,3-dihydrobenzo[b][1,4]dioxine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Methoxy-2,3-dihydro-1,4-benzodioxine

27593-500MG 27593-1G 27593-5G 27593-25G
77324-886EA 111.24 USD
77324-886 77343-288 77361-212 77372-976
6-Methoxy-2,3-dihydrobenzo[b][1,4]dioxine ≥97%
6-Methoxy-2,3-dihydrobenzo[b][1,4]dioxine

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14707737
CAS Number: 29668-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR