Methyl 2-fluoro-3,6-dimethoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

27572-500MG 27572-1G 27572-5G
77335-460EA 354.32 USD
77335-460 77348-724 77372-974
Methyl 2-fluoro-3,6-dimethoxybenzoate ≥95%
Methyl 2-fluoro-3,6-dimethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₄
MW: 214.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19439909
CAS Number: 1806390-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR