2-Fluoro-3,6-dimethoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

27571-500MG 27571-1G 27571-5G
77336-058EA 375.95 USD
77336-058 77348-722 77372-972
2-Fluoro-3,6-dimethoxybenzamide ≥97%
2-Fluoro-3,6-dimethoxybenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₃
Storage Temperature: Ambient
MDL Number: MFCD19439917
CAS Number: 1379077-97-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR