2-Fluoro-3,6-dimethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

27570-500MG 27570-1G 27570-5G 27570-25G
77324-884EA 242.05 USD
77324-884 77348-720 77361-210 77384-484
2-Fluoro-3,6-dimethoxybenzoic acid ≥97%
2-Fluoro-3,6-dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₄
Storage Temperature: Ambient
MDL Number: MFCD19439908
CAS Number: 1352306-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR