2-Fluoro-6-hydroxy-3-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

27568-500MG 27568-1G 27568-5G 27568-25G
77324-882EA 225.57 USD
77324-882 77348-718 77361-208 77384-482
2-Fluoro-6-hydroxy-3-methoxybenzoic acid ≥97%
2-Fluoro-6-hydroxy-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₄
Storage Temperature: Ambient
MDL Number: MFCD28554877
CAS Number: 133788-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR