1-(2-Bromo-6-fluoro-3-methoxyphenyl)-N-methylmethanamine ≥97%

Supplier: AOBChem USA

Warning

27498-500MG 27498-1G 27498-5G 27498-25G
77324-880EA 176.13 USD
77324-880 77346-518 77361-206 77384-480
1-(2-Bromo-6-fluoro-3-methoxyphenyl)-N-methylmethanamine ≥97%
1-(2-Bromo-6-fluoro-3-methoxyphenyl)-N-methylmethanamine

Precaución: for Research Use Only

Formula: C₉H₁₁BrFNO
MW: 248.092 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28142597
CAS Number: 1779900-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR