2,6-Dimethylbiphenyl-4-ol ≥95%

Supplier: AOBChem USA

Warning

27497-500MG 27497-1G 27497-5G
77335-640EA 361.53 USD
77335-640 77348-716 77372-970
2,6-Dimethylbiphenyl-4-ol ≥95%
2,6-Dimethylbiphenyl-4-ol

Precaución: for Research Use Only

Formula: C₁₄H₁₄O
MW: 198.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22627869
CAS Number: 100444-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR