1-Bromo-4-[[4-[(trifluoromethyl)oxy]phenyl]oxy]benzene ≥97%

Supplier: AOBChem USA

Warning

27387-500MG 27387-1G 27387-5G 27387-25G
77324-874EA 83.43 USD
77324-874 77342-188 77361-198 77372-960
1-Bromo-4-[[4-[(trifluoromethyl)oxy]phenyl]oxy]benzene ≥97%
1-Bromo-4-[[4-[(trifluoromethyl)oxy]phenyl]oxy]benzene

Precaución: for Research Use Only

Formula: C₁₃H₈BrF₃O₂
MW: 333.101 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30474680
CAS Number: 873203-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR